The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011716.2 | NP_035846.1 | 2 - 3 | 354 | 80 |
GenBank mRNA | AF084482.1 | - | 2 - 3 | 287 | 80 |
AJ011971.1 | - | 2 - 3 | 281 | 80 | |
AK144561.1 | - | 2 - 3 | 306 | 80 | |
BC046988.1 | - | 2 - 3 | 290 | 80 |