Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfr
Entrez Gene ID
22763
Gene Name
zinc finger RNA binding protein
Gene Aliases
C920030H05Rik
Chromosome Location
Chr.15: 12117793 - 12185449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011767.2 NP_035897.2 11 - 12 2034 79
GenBank mRNA AF071059.1 - 11 - 12 2032 79
AK034963.1 - 4 - 5 603 79
AK158938.1 - 3 - 4 342 79
BC006962.1 - 1 - 2 81 79
BC094220.1 - 10 - 11 1921 79
BC141139.1 - 11 - 12 2018 79

View Full Product Details