Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ikzf2
Entrez Gene ID
22779
Gene Name
IKAROS family zinc finger 2
Gene Aliases
A730095J18Rik, Helios, Zfpn1a2, Znfn1a2
Chromosome Location
Chr.1: 69531207 - 69687351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011770.4 NP_035900.2 5 - 6 856 76
GenBank mRNA AF044257.1 - 5 - 6 751 76
AK047925.1 - 5 - 6 718 76
BC138606.1 - 5 - 6 696 76
BC138608.1 - 5 - 6 696 76

View Full Product Details