Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gfpt2
Entrez Gene ID
14584
Gene Name
glutamine fructose-6-phosphate transaminase 2
Gene Aliases
AI480523, GFAT2
Chromosome Location
Chr.11: 49794155 - 49838620 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013529.3 NP_038557.1 8 - 9 761 81
GenBank mRNA AB016780.1 - 8 - 9 727 81
AK132821.1 - 7 - 8 607 81
BC031928.1 - 8 - 9 729 81

View Full Product Details