Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap14
Entrez Gene ID
23927
Gene Name
keratin associated protein 14
Gene Aliases
AA589630, Pmg1, mKAP13
Chromosome Location
Chr.16: 88825291 - 88826145 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013707.2 NP_038735.2 1 - 1 176 82
GenBank mRNA AF003691.1 - 1 - 1 168 82
BC104262.1 - 1 - 1 150 82
BC104263.1 - 1 - 1 149 82
CJ056567.1 - 1 - 1 177 82
D85925.1 - 1 - 1 165 82

View Full Product Details