The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013736.4 | NP_038764.2 | 3 - 4 | 261 | 83 |
GenBank mRNA | AB025015.1 | - | 3 - 4 | 245 | 83 |
AK036592.1 | - | 3 - 4 | 262 | 83 | |
AK145994.1 | - | 3 - 4 | 262 | 83 | |
AK147088.1 | - | 3 - 4 | 256 | 83 | |
BC049885.1 | - | 3 - 4 | 257 | 83 |