The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015790.3 | NP_056605.1 | 4 - 5 | 940 | 68 |
GenBank mRNA | AF199027.1 | - | 4 - 5 | 849 | 68 |
AF216747.1 | - | 3 - 4 | 783 | 68 | |
AF394451.1 | - | - | 783 | 68 | |
AK041320.1 | - | 3 - 4 | 1408 | 68 | |
AK041421.1 | - | 4 - 5 | 950 | 68 | |
AK220220.1 | - | 1 - 2 | 667 | 68 | |
BC029227.1 | - | 4 - 5 | 940 | 68 |