The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016808.2 | NP_058088.2 | 11 - 12 | 1796 | 71 |
NM_198091.2 | NP_932759.1 | 10 - 11 | 1252 | 71 | |
NM_198092.2 | NP_932760.2 | 11 - 12 | 1893 | 71 | |
GenBank mRNA | AB041799.1 | - | 10 - 11 | 1213 | 71 |
AF079565.1 | - | 10 - 11 | 854 | 71 | |
AK138799.1 | - | 12 - 13 | 2094 | 71 | |
AY255637.1 | - | 10 - 11 | 1221 | 71 | |
AY255638.1 | - | 11 - 12 | 1775 | 71 | |
AY255639.1 | - | 11 - 12 | 1729 | 71 | |
AY255640.1 | - | 11 - 12 | 1774 | 71 | |
BC017517.1 | - | 11 - 12 | 1775 | 71 |