The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018795.2 | NP_061265.2 | 15 - 16 | 1941 | 89 |
GenBank mRNA | AB028737.1 | - | 15 - 16 | 1947 | 89 |
BC040400.1 | - | 15 - 16 | 1949 | 89 | |
EF109740.1 | - | 15 - 16 | 1941 | 89 | |
EF109741.1 | - | 15 - 16 | 1941 | 89 |