Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcb10
Entrez Gene ID
56199
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 10
Gene Aliases
Abc-me, Abcb12
Chromosome Location
Chr.8: 123952459 - 123983122 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019552.2 NP_062425.1 2 - 3 917 58
GenBank mRNA AF266284.1 - 2 - 3 647 58
AK081782.1 - 2 - 3 629 58
BC046818.1 - 2 - 3 873 58
BC053020.1 - 2 - 3 873 58
BC054793.1 - 2 - 3 917 58

View Full Product Details