Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgm1
Entrez Gene ID
21816
Gene Name
transglutaminase 1, K polypeptide
Gene Aliases
2310004J08Rik, Tgase1
Chromosome Location
Chr.14: 55700009 - 55713921 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161714.1 NP_001155186.1 1 - 2 105 57
GenBank mRNA AF186373.1 - 1 - 2 96 57

View Full Product Details