Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmeb1
Entrez Gene ID
56809
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
1110050A04Rik, AI256615, AI481278
Chromosome Location
Chr.4: 132220376 - 132261549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122992.1 NP_001116464.1 6 - 7 585 101
NM_020273.2 NP_064669.2 5 - 6 522 101
GenBank mRNA AF210433.1 - 4 - 5 483 101
AK134349.1 - 5 - 6 523 101
BC046983.1 - 6 - 7 556 101
BU701415.1 - 4 - 5 369 101
DV660282.1 - 4 - 5 440 101

View Full Product Details