Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttyh1
Entrez Gene ID
57776
Gene Name
tweety homolog 1 (Drosophila)
Gene Aliases
4930459B04Rik, 6330408P11Rik, TTY1, mTTY1, tty
Chromosome Location
Chr.7: 4119527 - 4136241 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF190699.1 - 1 - 2 199 99
AF190991.1 - 1 - 2 199 99
AK018148.1 - 1 - 2 191 99
AK031550.1 - 1 - 2 227 99
AK042758.1 - 1 - 2 23 99
AK043998.1 - 1 - 2 247 99
AK048413.1 - 1 - 2 198 99
AK081981.1 - 1 - 2 200 99
BC046310.1 - 1 - 2 185 99
BC065694.1 - 1 - 2 191 99

View Full Product Details