The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277957.1 | NP_001264886.1 | 2 - 3 | 330 | 75 |
NM_001277958.1 | NP_001264887.1 | 2 - 3 | 298 | 75 | |
NM_001277959.1 | NP_001264888.1 | 2 - 3 | 298 | 75 | |
NM_021466.3 | NP_067441.1 | 2 - 3 | 330 | 75 | |
GenBank mRNA | AK040747.1 | - | 2 - 3 | 234 | 75 |
AK132811.1 | - | 2 - 3 | 284 | 75 | |
AK133426.1 | - | 2 - 3 | 299 | 75 | |
BC061106.1 | - | 2 - 3 | 215 | 75 | |
BC138425.1 | - | 2 - 3 | 196 | 75 | |
CN707961.1 | - | 2 - 3 | 330 | 75 | |
Y09972.1 | - | 2 - 3 | 232 | 75 |