Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wrap73
Entrez Gene ID
59002
Gene Name
WD repeat containing, antisense to Trp73
Gene Aliases
2610044M17Rik, 5330425N03Rik, Dd57, Wdr8
Chromosome Location
Chr.4: 154142357 - 154168259 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021499.2 NP_067474.2 1 - 2 144 101
GenBank mRNA AB034911.2 - 1 - 2 124 101
AK011773.1 - 1 - 2 108 101
AK051688.1 - 1 - 2 88 101
AK156969.1 - 1 - 2 143 101
AK160331.1 - 1 - 2 116 101
AK172690.1 - 1 - 2 143 101
BC016120.1 - 1 - 2 107 101
BY063723.1 - 1 - 2 145 101

View Full Product Details