The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021791.3 | NP_068563.2 | 10 - 11 | 1106 | 103 |
GenBank mRNA | AB046665.1 | - | 9 - 10 | 1104 | 103 |
BC061104.1 | - | 10 - 11 | 1106 | 103 |