Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oca2
Entrez Gene ID
18431
Gene Name
oculocutaneous albinism II
Gene Aliases
D7H15S12, D7Icr28RN, D7Nic1, p
Chromosome Location
Chr.7: 56239593 - 56536517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021879.2 NP_068679.1 18 - 19 2066 63
GenBank mRNA AK051897.1 - 17 - 18 1907 63
AK087494.1 - 19 - 20 2326 63
BC119220.1 - 18 - 19 2036 63
BC120549.1 - 18 - 19 2036 63
M97900.1 - 18 - 19 2066 63

View Full Product Details