Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pmel
Entrez Gene ID
20431
Gene Name
premelanosome protein
Gene Aliases
D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87
Chromosome Location
Chr.10: 128705168 - 128720238 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021882.4 NP_068682.2 8 - 9 1483 68
GenBank mRNA AF119092.1 - 3 - 4 360 68
AK012808.1 - 8 - 9 1484 68
AK131956.1 - 8 - 9 1485 68
BC075667.1 - 8 - 9 1468 68
BC082555.1 - 8 - 9 1469 68
U14133.1 - 8 - 9 1458 68

View Full Product Details