The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022028.2 | NP_071311.1 | 4 - 5 | 1188 | 77 |
GenBank mRNA | AJ292968.1 | - | 4 - 5 | 1188 | 77 |
AK006655.1 | - | 3 - 4 | 625 | 77 | |
AK034983.1 | - | 2 - 3 | 2061 | 77 | |
BC019377.1 | - | 4 - 5 | 1121 | 77 |