Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Oxct2a
- Entrez Gene ID
-
64059
- Gene Name
- 3-oxoacid CoA transferase 2A
- Gene Aliases
-
Oxct, Oxct2, Scot, Scot-t1
- Chromosome Location
-
Chr.4: 123321875 - 123323679 on Build GRCm38
- Gene Symbol
- Gm17244
- Entrez Gene ID
-
100038759
- Gene Name
- predicted gene, 17244
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK043708.1 |
- |
1 - 1 |
|
69 |
AK081449.1 |
- |
1 - 1 |
|
69 |
- Gene Symbol
- Oxct2b
- Entrez Gene ID
-
353371
- Gene Name
- 3-oxoacid CoA transferase 2B
- Gene Aliases
-
Scot-t2
- Chromosome Location
-
Chr.4: 123116248 - 123118000 on Build GRCm38
View Full Product Details