Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lpin3
Entrez Gene ID
64899
Gene Name
lipin 3
Gene Aliases
9130206L11Rik, AA438110, AV236139, mKIAA4023
Chromosome Location
Chr.2: 160880670 - 160906000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199118.1 NP_001186047.1 4 - 5 788 79
NM_022883.3 NP_075021.1 4 - 5 865 79
GenBank mRNA AF286724.1 - 4 - 5 802 79
AK033647.1 - 4 - 5 786 79
AK049363.1 - 4 - 5 781 79
AK163833.1 - 4 - 5 780 79
AK220228.1 - 4 - 5 865 79
BC026729.1 - 4 - 5 772 79
BC117882.1 - 3 - 4 729 79
BC117883.1 - 3 - 4 729 79

View Full Product Details