The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022993.3 | NP_075369.2 | 6 - 7 | 1982 | 75 |
GenBank mRNA | AB042200.1 | - | 6 - 7 | 1884 | 75 |
AK132169.1 | - | - | 1966 | 75 | |
AK144035.1 | - | 6 - 7 | 1995 | 75 | |
BC011058.1 | - | 2 - 3 | 262 | 75 | |
BC052378.1 | - | 6 - 7 | 1958 | 75 | |
EF437372.1 | - | 1 - 2 | 91 | 75 |