Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lcmt1
Entrez Gene ID
30949
Gene Name
leucine carboxyl methyltransferase 1
Gene Aliases
AL033290, LCMT-1, Lcmt
Chromosome Location
Chr.7: 123377803 - 123430358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025304.3 NP_079580.2 8 - 9 952 66
GenBank mRNA AK003424.1 - 8 - 9 877 66
AK007326.1 - 2 - 3 201 66
AK219030.1 - 1 - 2 56 66
BC010316.1 - 5 - 6 405 66
BC132507.1 - 8 - 9 858 66
BC132509.1 - 8 - 9 858 66
BC145301.1 - 8 - 9 858 66

View Full Product Details