The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025681.2 | NP_079957.2 | 3 - 4 | 648 | 63 |
GenBank mRNA | AF351204.1 | - | 3 - 4 | 389 | 63 |
AK012220.1 | - | 3 - 4 | 456 | 63 | |
AK017685.1 | - | 3 - 4 | 467 | 63 | |
BC049574.1 | - | 3 - 4 | 662 | 63 | |
BC052700.1 | - | 3 - 4 | 448 | 63 | |
BC063057.1 | - | 3 - 4 | 441 | 63 |