The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053191.2 | NP_444421.2 | 2 - 3 | 448 | 62 |
GenBank mRNA | AF329196.1 | - | 2 - 3 | 368 | 62 |
AK030960.1 | - | 2 - 3 | 449 | 62 | |
AK040961.1 | - | 2 - 3 | 447 | 62 | |
AK165584.1 | - | 1 - 2 | 124 | 62 | |
BC116705.1 | - | 2 - 3 | 344 | 62 |