The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080595.2 | NP_542162.1 | 5 - 6 | 463 | 67 |
GenBank mRNA | AJ416093.1 | - | 5 - 6 | 463 | 67 |
BC130023.1 | - | 5 - 6 | 468 | 67 |