Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pask
Entrez Gene ID
269224
Gene Name
PAS domain containing serine/threonine kinase
Gene Aliases
mKIAA0135
Chromosome Location
Chr.1: 93309437 - 93343350 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080850.2 NP_543126.2 4 - 5 910 91
GenBank mRNA AK028435.1 - 4 - 5 912 91
AK083528.1 - 4 - 5 881 91
AK083590.1 - 4 - 5 875 91
AK129063.1 - 4 - 5 800 91
AK172031.1 - 4 - 5 858 91
BC150821.1 - 3 - 4 622 91

View Full Product Details