Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh18
Entrez Gene ID
73173
Gene Name
protocadherin 18
Gene Aliases
3110038E07Rik, BB095589, PCDH68L
Chromosome Location
Chr.3: 49743291 - 49757382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130448.3 NP_569715.3 1 - 2 2946 92
GenBank mRNA AF416735.1 - 1 - 2 2891 92
AK014140.1 - 1 - 2 1094 92
AK036560.1 - 1 - 2 2915 92
AK173204.1 - 1 - 2 2891 92
BC052198.1 - 1 - 2 2910 92

View Full Product Details