The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130452.1 | NP_569719.1 | 6 - 7 | 1095 | 109 |
GenBank mRNA | AK050010.1 | - | - | 1113 | 109 |
AY033514.1 | - | 6 - 7 | 1033 | 109 | |
BC019406.1 | - | 6 - 7 | 1095 | 109 |