Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nphs2
Entrez Gene ID
170484
Gene Name
nephrosis 2, podocin
Gene Aliases
AI790225, PDCN, SRN1
Chromosome Location
Chr.1: 156310544 - 156328035 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130456.3 NP_569723.1 3 - 4 514 129
GenBank mRNA AJ302048.1 - 3 - 4 461 129
AK052746.1 - 3 - 4 523 129
AY050309.1 - 3 - 4 537 129
BC067401.1 - 3 - 4 514 129
FJ715770.1 - 3 - 4 522 129
FJ715771.1 - 3 - 4 552 129
FJ715772.1 - 3 - 4 552 129
FJ715773.1 - 3 - 4 552 129
FJ715774.1 - 3 - 4 552 129
FJ715775.1 - 3 - 4 552 129
FJ715776.1 - 3 - 4 552 129

View Full Product Details