Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk9
Entrez Gene ID
107951
Gene Name
cyclin-dependent kinase 9 (CDC2-related kinase)
Gene Aliases
PITALRE
Chromosome Location
Chr.2: 32705782 - 32712784 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130860.3 NP_570930.1 5 - 6 735 67
GenBank mRNA AF327431.1 - 5 - 6 607 67
AK089276.1 - 5 - 6 574 67
AK142397.1 - 3 - 4 770 67
AK143217.1 - 5 - 6 736 67
AK144607.1 - 5 - 6 716 67
AK157340.1 - 5 - 6 707 67
BC003901.1 - 5 - 6 680 67

View Full Product Details