Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdhr1
Entrez Gene ID
170677
Gene Name
cadherin-related family member 1
Gene Aliases
Pcdh21, Prcad, mKIAA1775
Chromosome Location
Chr.14: 37077849 - 37098311 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130878.2 NP_570948.1 15 - 16 1951 77
GenBank mRNA AB093300.1 - 15 - 16 2367 77
AF426393.1 - 15 - 16 1838 77
AK143151.1 - 15 - 16 1892 77
BC042454.1 - 15 - 16 1861 77
BC058270.1 - 15 - 16 1951 77

View Full Product Details