Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Api5
Entrez Gene ID
11800
Gene Name
apoptosis inhibitor 5
Gene Aliases
AAC-11, AI196452, API-5
Chromosome Location
Chr.2: 94411727 - 94438181 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK134480.1 - 9 - 10 1255 73
AK137466.1 - 9 - 10 1284 73
AK153934.1 - 9 - 10 1228 73
AK160874.1 - 9 - 10 1252 73
AK167315.1 - 9 - 10 1227 73
AK167375.1 - 9 - 10 1268 73
BC007133.1 - 9 - 10 1247 73
U35846.1 - 9 - 10 1212 73

View Full Product Details