The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077265.2 | NP_001070733.1 | 1 - 2 | 542 | 74 |
NM_007516.3 | NP_031542.2 | 1 - 2 | 542 | 74 | |
GenBank mRNA | AK135733.1 | - | 1 - 2 | 488 | 74 |
BC011172.1 | - | 1 - 2 | 217 | 74 | |
BC049098.1 | - | 1 - 2 | 542 | 74 |