The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007608.2 | NP_031634.2 | 1 - 2 | 242 | 87 |
GenBank mRNA | AI098626.1 | - | 1 - 2 | 158 | 87 |
AI314393.1 | - | 1 - 2 | 242 | 87 | |
AI314492.1 | - | 1 - 2 | 224 | 87 | |
BC030174.1 | - | 1 - 2 | 220 | 87 | |
X51971.1 | - | 1 - 2 | 234 | 87 |