The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008485.3 | NP_032511.3 | 5 - 6 | 814 | 69 |
GenBank mRNA | U43327.2 | - | 4 - 5 | 547 | 69 |