The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008779.2 | NP_032805.2 | 21 - 22 | 3065 | 64 |
GenBank mRNA | AK140747.1 | - | 12 - 13 | 1654 | 64 |
L01991.1 | - | - | 3174 | 64 |