Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfx1
Entrez Gene ID
19724
Gene Name
regulatory factor X, 1 (influences HLA class II expression)
Gene Aliases
AI047719, AI385641
Chromosome Location
Chr.8: 84066675 - 84096992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009055.4 NP_033081.3 14 - 15 2167 65
GenBank mRNA AK037226.1 - 7 - 8 1110 65
AK083571.1 - 14 - 15 2138 65
AK147790.1 - 14 - 15 2172 65
BC057018.1 - 14 - 15 2146 65
X76088.1 - 14 - 15 2074 65

View Full Product Details