The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009061.4 | NP_033087.2 | 1 - 2 | 134 | 73 |
GenBank mRNA | AF215668.1 | - | 1 - 2 | 119 | 73 |
AF432916.1 | - | 1 - 2 | 113 | 73 | |
AK029310.1 | - | 1 - 2 | 136 | 73 | |
AK031603.1 | - | 1 - 2 | 136 | 73 | |
AK077922.1 | - | 1 - 2 | 135 | 73 | |
AK162276.1 | - | 1 - 2 | 136 | 73 | |
BC023001.1 | - | 1 - 2 | 117 | 73 | |
U67187.1 | - | 1 - 2 | 123 | 73 |