The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205321.1 | NP_001192250.1 | 3 - 4 | 465 | 82 |
NM_009134.3 | NP_033160.2 | 3 - 4 | 465 | 82 | |
GenBank mRNA | AK157866.1 | - | 3 - 4 | 465 | 82 |
AY538273.1 | - | 2 - 3 | 395 | 82 | |
Y09108.1 | - | 2 - 3 | 430 | 82 |