The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010158.2 | NP_034288.2 | 5 - 6 | 938 | 66 |
GenBank mRNA | AF079763.1 | - | 5 - 6 | 946 | 66 |
AF099092.1 | - | 5 - 6 | 949 | 66 | |
AK132792.1 | - | 5 - 6 | 639 | 66 | |
AK160564.1 | - | 2 - 3 | 285 | 66 | |
BC031507.1 | - | 5 - 6 | 684 | 66 | |
BC057577.1 | - | 5 - 6 | 922 | 66 |