Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glra4
Entrez Gene ID
14657
Gene Name
glycine receptor, alpha 4 subunit
Gene Aliases
-
Chromosome Location
Chr.X: 136756628 - 136792158 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010297.2 NP_034427.2 3 - 4 287 61
GenBank mRNA AF462147.1 - 2 - 3 149 61
BC110630.2 - 2 - 3 149 61
DQ109809.1 - 3 - 4 287 61

View Full Product Details