The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010297.2 | NP_034427.2 | 6 - 7 | 730 | 88 |
GenBank mRNA | AF462147.1 | - | 5 - 6 | 592 | 88 |
AK085727.1 | - | 6 - 7 | 1153 | 88 | |
BC110630.2 | - | 5 - 6 | 592 | 88 | |
DQ109809.1 | - | 6 - 7 | 730 | 88 |