Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tes
Entrez Gene ID
21753
Gene Name
testis derived transcript
Gene Aliases
D6Ertd352e, TESS, Tes1, Tes2, testin2
Chromosome Location
Chr.6: 17065149 - 17105825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207176.3 NP_997059.1 3 - 4 508 104
GenBank mRNA AK132249.1 - 3 - 4 466 104
AK145609.1 - 3 - 4 457 104
AK167101.1 - 3 - 4 457 104
AK167274.1 - 3 - 4 465 104
BC003808.1 - 5 - 6 748 104
BC010465.1 - 3 - 4 464 104
BC051103.1 - 3 - 4 442 104
CT010319.1 - 2 - 3 343 104
X78989.1 - 2 - 3 340 104

View Full Product Details