Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx5a
Entrez Gene ID
56389
Gene Name
syntaxin 5A
Gene Aliases
0610031F24Rik, D19Ertd627e, Stx5
Chromosome Location
Chr.19: 8741424 - 8755642 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167799.1 NP_001161271.1 8 - 9 853 77
NM_019829.4 NP_062803.4 7 - 8 1152 77
GenBank mRNA AF232709.1 - 7 - 8 520 77
AK166087.1 - 7 - 8 1155 77
AK190896.1 - 3 - 4 187 77
AK201004.1 - 2 - 3 282 77
BC004849.1 - 8 - 9 786 77
BC021883.1 - 7 - 8 1135 77
DV649852.1 - 2 - 3 116 77

View Full Product Details