Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tssc4
Entrez Gene ID
56844
Gene Name
tumor-suppressing subchromosomal transferable fragment 4
Gene Aliases
AA241958, ESTM671070
Chromosome Location
Chr.7: 143068035 - 143071093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020285.2 NP_064681.1 2 - 3 244 60
NM_138631.1 NP_619537.1 2 - 3 244 60
GenBank mRNA AB038011.1 - 2 - 3 244 60
AJ279796.1 - 2 - 3 244 60
AJ279797.1 - 2 - 3 244 60
AK088052.1 - 2 - 3 179 60

View Full Product Details