The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023275.2 | NP_075764.1 | 1 - 2 | 685 | 80 |
GenBank mRNA | AB060651.1 | - | 1 - 2 | 590 | 80 |
AF309564.1 | - | 1 - 2 | 544 | 80 | |
AJ276568.1 | - | 1 - 2 | 144 | 80 | |
AK003482.1 | - | 1 - 2 | 573 | 80 | |
AK003490.1 | - | 1 - 2 | 573 | 80 | |
AK087019.1 | - | 1 - 2 | 326 | 80 | |
AK156619.1 | - | 1 - 2 | 573 | 80 | |
AK157518.1 | - | 1 - 2 | 573 | 80 | |
AK159385.1 | - | 1 - 2 | 606 | 80 | |
BC024626.1 | - | 1 - 2 | 565 | 80 | |
BC043719.1 | - | 1 - 2 | 622 | 80 | |
BC054464.1 | - | 1 - 2 | 674 | 80 | |
BC060193.1 | - | 1 - 2 | 563 | 80 |