The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025385.3 | NP_079661.1 | 1 - 2 | 90 | 109 |
GenBank mRNA | AK003850.1 | - | 1 - 2 | 87 | 109 |
BY285374.1 | - | 1 - 2 | 92 | 109 |