The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025388.2 | NP_079664.1 | 6 - 7 | 562 | 96 |
GenBank mRNA | AK003070.1 | - | 5 - 6 | 464 | 96 |
AK003897.1 | - | 6 - 7 | 563 | 96 | |
AK011260.1 | - | 5 - 6 | 453 | 96 | |
BC021936.1 | - | 6 - 7 | 523 | 96 | |
BC055109.1 | - | 1 - 2 | 1353 | 96 |