Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mien1
Entrez Gene ID
103742
Gene Name
migration and invasion enhancer 1
Gene Aliases
1810046J19Rik, AI463380, Rdx12
Chromosome Location
Chr.11: 98437708 - 98438988 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025559.2 NP_079835.1 3 - 4 307 70
GenBank mRNA AK002959.1 - 3 - 4 290 70
AK007795.1 - 3 - 4 290 70
AK009922.1 - 3 - 4 285 70
AK041314.1 - 3 - 4 289 70
BC021589.1 - 3 - 4 291 70
CA530220.1 - 2 - 3 133 70

View Full Product Details